missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMCD1 Antibody (OTI1H5), Allophycocyanin/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71865APCCY7
This item is not returnable.
View return policy
Description
LMCD1 Monoclonal specifically detects LMCD1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| LMCD1 | |
| Monoclonal | |
| APC-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| LMCD1 | |
| Full length recombinant protein of human LMCD1 (NP_055398) produced in HEK293T cells. | |
| 0.1 mL | |
| 29995.0 | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| OTI1H5 | |
| Flow Cytometry | |
| dyxin, LIM and cysteine-rich domains 1, LIM and cysteine-rich domains protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction