missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMCD1 Mouse anti-Human, Clone: OTI1H5, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503139
This item is not returnable.
View return policy
Description
LMCD1 encodes a member of the LIM-domain family of zinc finger proteins. The encoded protein contains an N-terminal cysteine-rich domain and two C-terminal LIM domains. The presence of LIM domains suggests involvement in protein-protein interactions. The protein may act as a co-regulator of transcription along with other transcription factors. Alternate splicing results in multiple transcript variants of this gene.Specifications
| LMCD1 | |
| Monoclonal | |
| 0.8 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZU5 | |
| LMCD1 | |
| Full length human recombit protein of human LMCD1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H5 | |
| Unconjugated | |
| LMCD1 | |
| LIM and cysteine rich domains 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 29995 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction