missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals Rat LIFR alpha ELISA Kit (Colorimetric)

Product Code. 18663400 Shop All Bio Techne Products
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 18663400

Brand: Novus Biologicals NBP280386

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Assay Range 46.88-3000 pg/mL
Assay Sensitivity 13 pg/mL
Product Type ELISA Kit (Colorimetric)
Sample Type Serum
Specificity Recognizes both recombinant and natural Rat LIFR alpha.
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3977
Gene Symbol LIFR
Quantity 1 Kit
Storage Requirements Storage is content dependent.
Synonym CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS
Target Rat
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.