missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LMO1 Polyclonal Antibody

Description
Recommended positive controls: 293T, mouse brain, rat brain. Predicted reactivity: Mouse (98%), Rat (99%), Zebrafish (98%), Xenopus laevis (98%), Pig (96%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
LMO1 encodes a cysteine-rich, two LIM domain transcriptional regulator. It is mapped to an area of consistent chromosomal translocation in chromosome 11, disrupting it in T-cell leukemia, although more rarely than the related gene, LMO2 is disrupted.
Specifications
Specifications
| Antigen | LMO1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.87 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | LMO1 |
| Gene Accession No. | P25800, Q924W9 |
| Gene Alias | cb524; Cysteine-rich protein TTG-1; Dat1; dopamine-inducible LIM-domain transcription factor DAT1; etID41074.7; LIM domain only 1; LIM domain only 1 (rhombotin 1); LIM domain only 3; LIM domain only protein 1; LIM domain only protein 3; LIM-only 1; Lmo1; LMO-1; lmo1b; Lmo3; LMO-3; neuronal-specific transcription factor DAT1; RBTN1; Rbtn-1; RHOM1; rhombotin 1; rhombotin-1; T-cell translocation gene 1; T-cell translocation protein 1; transcription factor; Ttg1; Unknown (protein for MGC:142594); wu:fb76g02; zgc:109794 |
| Gene Symbols | LMO1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?