missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMO2 Antibody (OTI1G10), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71157APC
This item is not returnable.
View return policy
Description
LMO2 Monoclonal antibody specifically detects LMO2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| LMO2 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI1G10 | |
| Western Blot, Immunohistochemistry | |
| Cysteine-rich protein TTG-2, LIM domain only 2 (rhombotin-like 1), LIM domain only protein 2, LMO-2, RBTN2rhombotin-2, RBTNL1rhombotin-like 1, RHOM2T-cell translocation protein 2, T-cell translocation gene 2, TTG2rhombotin 2 | |
| Full length human recombinant protein of human LMO2(NP_005565) produced in HEK293T cell. | |
| 0.1 mL | |
| Angiogenesis, Cancer | |
| 4005 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction