missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMO2 Monoclonal Antibody (OTI1D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506133S
This item is not returnable.
View return policy
Description
LMO2 Monoclonal antibody specifically detects LMO2 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| LMO2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P25791 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4005 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D7 | |
| Unconjugated | |
| Lmo2 | |
| cysteine-rich protein TTG-2; LIM domain only 2; LIM domain only 2 (rhombotin-like 1); LIM domain only protein 2; LIM domain only-2; LIM only 2; LMO2; LMO-2; LMO2 protein; Rbtn2; Rbtn-2; RBTNL1; RHOM2; Rhom-2; rhombotin 2; rhombotin-2; rhombotin-like 1; si:dkey-10o6.5; T-cell translocation gene 2; T-cell translocation protein 2; TTG2; wu:fc83a08; zgc:111930; zlmo2 | |
| Full length human recombinant protein of human LMO2 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction