missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LMO4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15771742 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15771742 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15771742 Supplier Invitrogen™ Supplier No. PA563976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FTCSTCRNRL VPGDRFHYIN GSLFCEHDRP TALINGHLNS LQSNPLLPDQ KVC Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

LMO4 encodes a cysteine-rich protein that contains two LIM domains but lacks a DNA-binding homeodomain. The encoded protein may play a role as a transcriptional regulator or as an oncogene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LMO4
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene LMO4
Gene Accession No. P61968
Gene Alias A730077C12Rik; Breast tumor autoantigen; Crp3; ethanol induced 4; Etohi4; LIM domain only 4; LIM domain only protein 4; LIM domain transcription factor LMO4; LIM-only 4 protein; LMO4; LMO-4
Gene Symbols LMO4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human LMO4. Recombinant protein control fragment (Product #RP-104064).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8543
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.