Learn More
LMP7/PSMB8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-33540
Product Code. 18787853
  • € 555.00 / 0.10mL
Description
LMP7/PSMB8 Polyclonal specifically detects LMP7/PSMB8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PSMB8