Learn More
LMTK2 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-32182IR
Product Code. 18786201
  • € 633.00 / 0.10mL
Description
LMTK2 Polyclonal antibody specifically detects LMTK2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LMTK2
Western Blot
Primary
Purified
Western Blot
The immunogen this antibody was made to, maps to a region between residue 675 to 725 of human Lemur Tyrosine Kinase 2 using the numbering given in entry NP_055731.2 (GeneID 22853).
Store at 4°C in the dark.