missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LMTK3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-62711-25ul
This item is not returnable.
View return policy
Description
LMTK3 Polyclonal antibody specifically detects LMTK3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| LMTK3 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| AATYK3, EC 2.7.11.1, lemur tyrosine kinase 3KIAA1883TYKLM3LMR3, serine/threonine-protein kinase LMTK3 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: CSCLPLERGDAVAGWGGHPALGCPHPPEDDSSLRAERGSLADLPMAPPASAPPEFLDP | |
| 25 μL | |
| Protein Kinase | |
| 114783 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction