missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LNK Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575638
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the SH2B adaptor family of proteins, which are involved in a range of signaling activities by growth factor and cytokine receptors. The encoded protein is a key negative regulator of cytokine signaling and plays a critical role in hematopoiesis. Mutations in this gene have been associated with susceptibility to celiac disease type 13 and susceptibility to insulin-dependent diabetes mellitus.
Specifications
| LNK | |
| Polyclonal | |
| Unconjugated | |
| SH2B3 | |
| AI429800; IDDM20; linker of T-cell receptor pathways; LNK; Lymphocyte adapter protein; lymphocyte-specific adapter protein Lnk; SH2B adapter protein 3; SH2B adaptor protein 3; Sh2b3; Signal transduction protein Lnk | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10019, 16923, 58838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.2 | |
| O09039, P50745, Q9UQQ2 | |
| SH2B3 | |
| Synthetic peptide corresponding to residues in Human LNK. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction