missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lnx1 Antibody (OTI1H9), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72517F
This item is not returnable.
View return policy
Description
Lnx1 Monoclonal antibody specifically detects Lnx1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| Lnx1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| OTI1H9 | |
| Western Blot | |
| EC 6.3.2.-, ligand of numb-protein X 1ligand of numb-protein X, LNXmulti-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX, MPDZ, Numb-binding protein 1, PDZ domain-containing ring finger protein 2, PDZRN2E3 ubiquitin-protein ligase LNX | |
| Human recombinant protein fragment corresponding to amino acids 260-528 of human LNX1 (NP_116011) produced in E.coli. | |
| 0.1 mL | |
| Cancer, Hypoxia, Lipid and Metabolism, Zinc Finger | |
| 84708 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction