missing translation for 'onlineSavingsMsg'
Learn More

LNX1 Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene

Product Code. 17558892
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17558892 30 μL 30µL
17538892 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17558892 Supplier Origene Technologies Supplier No. TA800747S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

LNX1 Monoclonal antibody specifically detects LNX1 in Human samples. It is validated for Western Blot

Specifications

Antigen LNX1
Applications Western Blot
Classification Monoclonal
Clone OTI1H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MGMT
Gene Accession No. P16455
Gene Alias E3 ubiquitin-protein ligase LNX; Ligand of Numb protein X 1; ligand of Numb-binding protein 1; Ligand of Numb-protein X 1; ligand of numb-protein X 1, E3 ubiquitin protein ligase; LNX; Lnx1; MPDZ; multi-PDZ-domain-containing protein, E3 ubiquitin-protein ligase LNX; numb-binding protein 1; PDZ domain-containing RING finger protein 2; PDZRN2; RING-type E3 ubiquitin transferase LNX; UNQ574/PRO1136
Host Species Mouse
Immunogen Full length human recombinant protein of human MGMT produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4255
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.