Learn More
Brannan™ Lo-Tox™ Mercury-Free Blue Spirit Filled Thermometers
A general purpose/stirring thermometer with a low toxic, environmentally friendly blue filling. Brannan™ Lo-Tox™ Mercury-Free Blue Spirit Filled Thermometers are each manufactured to traceable standards.
€ 5.20 - € 21.45
Specifications
Bottle Fill Blue Spirit
For Use With (Application) General use
Immersion Type Partial Immersion
Includes Brannan non roll end cap
Material Glass
View More Specs
Products 6
Product Code Brand Temperature Range (Metric) Subdivision Price Quantity & Availability  
Product Code Brand Temperature Range (Metric) Subdivision Price Quantity & Availability  
13098583
View Documents Promotion Details
Brannan™
44/801/8
-35°C to +50°C 1°C
€ 6.15
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
13028593
View Documents Promotion Details
Brannan™
44/803/8
-20°C to +150°C 1°C
€ 5.20
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
13038583
View Documents Promotion Details
Brannan™
44/805/8
-10°C to +210°C 1°C
€ 21.45
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-05-2024
to see stock.
Add to basket
13038603
View Documents Promotion Details
Brannan™
44/820/0
-1°C to +51° C 0.1°C
€ 16.60
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
13048603
View Documents Promotion Details
Brannan™
44/822/0
-1°C to +51° C 0.2°C
€ 15.60
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
13058603
View Documents Promotion Details
Brannan™
44/824/0
-10°C to +101°C 0.2°C
€ 17.95
Each
View alternative products
Save Up to 
»
Estimated Shipment: 23-05-2024
to see stock.
Add to basket
Description
Description
A general purpose/stirring thermometer with a low toxic, environmentally friendly blue filling. Brannan™ Lo-Tox™ Mercury-Free Blue Spirit Filled Thermometers are each manufactured to traceable standards.
  • Easy to read against white background
  • With safety suspension ring
  • With a blue non-roll top
  • Available in different temperature ranges
  • Available for either 76mm or total immersion
Specifications
Specifications
Blue Spirit
Partial Immersion
Glass
76 mm
General use
Brannan non roll end cap
Laboratory Thermometer