missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOC281370, Mouse anti-Bovine, ATTO390, Clone: 6C11-B3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB18106.200ug
This item is not returnable.
View return policy
Specifications
| LOC281370 | |
| Monoclonal | |
| Atto 390 | |
| In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide). | |
| - | |
| Mouse | |
| 200 μg | |
| Primary | |
| Bovine, Human | |
| Antibody | |
| IgG2a κ |
| ELISA, Immunocytochemistry, Immunofluorescence, Western Blot | |
| 6C11-B3 | |
| ELISA Immunocytochemistry (1:100) Immunofluorescence (1:100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| polyubiquitin | |
| LOC281370 | |
| Native bovine ubiquitin, conjugated to KLH. | |
| RUO | |
| 281370 | |
| Store at -20°C. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction