Learn More
Thermo Scientific Chemicals Locust bean gum
Product Code. p-7214756 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
Product Code. 17271262
Brand: Thermo Scientific Chemicals 466670010
Request Bulk or Custom Format
CAS: 9000-40-2
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 9000-40-2
Specifications
Quantity 1 kg
Chemical Name or Material Locust bean gum