missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lofepramine Hydrochloride, Mikromol™
Description
Lofepramine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Lofepramine Hydrochloride |
| CAS | 26786-32-3 |
| Analyte or Component Names | Lofepramine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26 H27 Cl N2 O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ethanone, 1-(4-chlorophenyl)-2-[[3-(10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl]methylamino]-, hydrochloride (1:1), Acetophenone, 4'-chloro-2-[[3-(10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl]methylamino]-, monohydrochloride (8CI), Ethanone, 1-(4-chlorophenyl)-2-[[3-(10,11-dihydro-5H-dibenz[b,f]azepin-5-yl)propyl]methylamino]-, monohydrochloride (9CI), Gamanil, Gamonil, Leo 640 hydrochloride, Lofepramine hydrochloride, Lopramine hydrochloride, Timelit, Tymelyt, WHR 2908A |
| InChI Formula | InChI=1S/C26H27ClN2O.ClH/c1-28(19-26(30)22-13-15-23(27)16-14-22)17-6-18-29-24-9-4-2-7-20(24)11-12-21-8-3-5-10-25(21)29;/h2-5,7-10,13-16H,6,11-12,17-19H2,1H3;1H |
| SMILES | Cl.CN(CCCN1c2ccccc2CCc3ccccc13)CC(=O)c4ccc(Cl)cc4 |
| IUPAC Name | 1-(4-chlorophenyl)-2-[3-(5,6-dihydrobenzo[b][1]benzazepin-11-yl)propyl-methylamino]ethanone;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?