Learn More
Invitrogen™ Lonigutamab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559470
 View more versions of this product
Product Code. 30284277
  • € 1204.00 / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lonigutamab is a monoclonal antibody that targets and binds to the insulin-like growth factor 1 receptor (IGF-1R), inhibiting its activity and thereby blocking the IGF-1R signaling pathway.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lonigutamab Humanized
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A