Learn More
Lovibond™ Aqua Comparitor Color Discs: pH
For testing water with the Aqua Comparator
Brand:  Lovibond™ 146120
Additional Details : Weight : 0.02800kg
 View more versions of this product
Product Code. 11735448
  • € 22.60 / Each
Estimated Shipment
02-03-2021
Add to basket
Description
Description
Economical replacement discs make it easy to test for pH using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
Specifications
6 to 7.6 pH
Aqua comparator™
Testing water with the Aqua comparator™
pH Bromthymol Blue Color Disc