Learn More
Lovibond™ Battery Pack for SpectroDirect Spectrophotometer
External 12 VDC, NiCd rechargeable battery pack for SpectroDirect spectrophotometer.
Brand: Lovibond™ 711050
Additional Details : Weight : 7.14000kg
Description
- Type: External NiCd rechargeable
- Voltage: 12VDC
Specifications
External Battery Pack | |
SP600 spectrophotometer |