Learn More
Lovibond™ MC500 Colorimeter Replacement Reagents: Phosphate, Ortho LR
For use with MC500 multi-parameter colorimeter
Brand: Lovibond™ 517651BT
Additional Details : Weight : 0.30000kg
Includes:
- Phosphate No. 1 LR Tablets, 100 pack
- Phosphate No. 2 LR Tablets, 100 pack
Description
For use with MC500 multi-parameter colorimeter
- Parameter: Phosphate LR, Ortho
- Method: Phosphomolybdic Acid/Ascorbic Acid
Specifications
Tablet | |
MC500 multi-parameter colorimeter | |
100 | |
Phosphomolybdic Acid/Ascorbic Acid |
Water quality testing | |
Phosphate No. 1 LR tablets (100 pack), phosphate No. 2 LR tablets (100 pack) | |
0.05 to 4mg/L | |
LR Phosphate, Ortho, Reagent Set |
Safety and Handling
- COMBI-PACK LR PHOSPHATE NO. 1 LR + NO. 2 LR
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H318-Causes serious eye damage.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P313-Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : sodium hydrogen sulphate, adipic acid, disodium disulphate