Learn More
Kautex™ LDPE Wide Mouth Bottles without Closures
Product Code. 10442335
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
300 mL
500 mL
750 mL
1000 mL
1500 mL
Unit Size:
Each
Pack of 144
Pack of 360
Pack of 46
Pack of 64
Product Code. 10442335
missing translation for 'mfr': Kautex™ 2000070535
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed with a wide opening for easy filling and cleaning. Kautex™ Wide Mouth Bottles are ideal for liquids, pastes and granules.
  • Manufactured from low density polyehtylene (LDPE) for flexibility and toughness
  • Natural translucent color, round bottles
  • Ideal for dispensing, sending samples, adhesives, transportation and storage 
  • Closures available separately
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Form Wide Neck
Material LDPE
Quantity 1Each
Closure Included No
Graduated No
Shape Round
Product Type Wide Neck Bottle