Learn More
Kautex™ LDPE Wide Mouth Bottles without Closures
Click to view available options
Capacity (Metric):
50 mL
100 mL
200 mL
300 mL
500 mL
750 mL
1000 mL
1500 mL
Unit Size:
Each
Pack of 144
Pack of 360
Pack of 46
Pack of 64
Description
- Manufactured from low density polyehtylene (LDPE) for flexibility and toughness
- Natural translucent color, round bottles
- Ideal for dispensing, sending samples, adhesives, transportation and storage
- Closures available separately
Specifications
Specifications
| Capacity (Metric) | 500 mL |
| Form | Wide Neck |
| Material | LDPE |
| Quantity | 1Each |
| Closure Included | No |
| Graduated | No |
| Shape | Round |
| Product Type | Wide Neck Bottle |