Learn More
Thermo Scientific™ Low DNA Binding Snap Cap Microcentrifuge Tube
Shop All Thermo Scientific Products
Description
The importance of low DNA binding microcentrifuge tubes cannot be overstated. These tubes are specifically engineered to minimize the adhesion of DNA to the tube walls, which is crucial in preserving the quantity and quality of DNA samples during various laboratory procedures.
Ensuring that maximum DNA recovery is achieved and reducing the risk of sample loss and contamination. Particularly vital in applications such as PCR, sequencing, and cloning, where even small losses can significantly impact the results.
Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
Additionally, the assurance that these tubes are free from PCR inhibitors, UV leachables, and human DNA further enhances the reliability of experimental outcomes by preventing any potential interference with sensitive assays.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | Natural |
| Material | Polypropylene Resin |
| Volume (Metric) | 2 mL |
| Sterility | Non-sterile |
| Graduated | Yes |
| Closure Type | Snap Cap |
| Fit for Purpose | PCR Ready |
| Bottom Shape | Conical |
| Green Features | ACT Ecolabel (Silver) |
| Show More |