Learn More
BRAND™ Low Form Borosilicate Glass Beakers
Shop All Brand Products
missing translation for 'orderingAttributeHoverText'
Capacity (Metric):
25 mL
100 mL
150 mL
250 mL
400 mL
600 mL
800 mL
1000 mL
3000 mL
missing translation for 'unitSize'
Each
Pack of 10
Pack of 4
Description
- Borisilicate glass construction (Boro 3.3)
- Conforms to DIN 12331 / ISO 3819 standards
- Includes graduations in white
- Integrated spout provides easy, spill-proof pouring
Spécification
Spécification
| Diameter (Metric) Outer | 100 mm |
| Capacity (Metric) | 800 mL |
| Material | Borosilicate Glass |
| Height (Metric) | 135 mm |
| Includes | Integrated Spout |
| Quantity | 10/Pk. |
| Type | Beaker |
| Graduated | Yes |
| Certifications/Compliance | ISO 3819, DIN 12331 |
| Style | Low Form |