Learn More
SI Analytics™ Low Maintenance pH Combination Electrode with Screw Plug Head
Shop All SI Analytics Products
Click to view available options
Length (Metric):
120 mm
225 mm
320 mm
425 mm
Junction:
Ceramic Diaphragm
KPG Annular Gap Diaphragm
Unit Size:
Each
Description
- PH combination electrode, type H8281HD, length 120 mm, D = 12 mm, 0 ... + 100 ° C
- Low Maintenance pH Combination Electrode with Screw Plug Head
- Low maintenance, glass shaft, KPG annular gap junction
- Referid™-electrolyte, Silamid™-reference system, dome membrane, H-glass, plug head, length 120 mm, 12 mm Ø, 0...+100 °C, 2...13 pH
Specifications
Specifications
| Connector Types | Screw Plug Head |
| Junction | KPG Annular Gap Diaphragm |
| Material | H-Glass |
| pH Range | 2 to 13 |
| Type | Low Maintenance pH Combination Electrode |
| Internal Reference | Silamid™ |
| Membrane | Dome |
| Model | H 8281 Hd |
| Temperature (Metric) Range | 0°C to 100°C |
| Length (Metric) | 120 mm |
| Show More |