missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VistaLab™ Technologies Pipette Tips: Low Retention
For use with Ovation and MLA Brand pipetters
Brand: VistaLab™ Technologies 4070-3332LR
Additional Details : Weight : 0.00100kg
Description
- Sterile, Low Retention tips packaged in VistaRak: 5 racks of 192 tips each
- Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, etc.
- Use VistaLab pipette tips to maximize the exceptional performance and benefits of Ovation pipettes
- All VistaLab pipettes are calibrated with VistaLab tips to provide a perfect fit and deliver reliable performance every time
Specifications
Low Retention | |
Filtered | |
Clear | |
Not Autoclavable | |
5 in. | |
Ideal for detergents, enzymes, viscous samples, foaming liquids, plasma, TRIS, BSA and Tween, PCR or other biological applications | |
Rack | |
VistaRak: 5 racks | |
Single-use | |
No |
1250 μL | |
Sterile | |
Blue | |
3.4 in. | |
Certified RNase/DNase free, non-pyrogenic | |
e-beam | |
Low Retention, Straight | |
192 Tips/Rack | |
MLA, Ovation | |
Polypropylene, Polyethylene (Filter) |