Learn More
Miele Professional™ Lower Basket U 874/1
Product Code. 10242981 Shop All Miele Products
missing translation for 'orderingAttributeHoverText'
:
Dimensions (D x W x H): 515 x 534 x 50mm
Product Code. 10242981
missing translation for 'mfr': Miele Professional™ 6442940
Only null left
Add to basket
Add to basket
Lower basket U 874/1 for the optimum loading of different inserts and mesh trays
  • Vertical clearance can vary with different upper baskets
  • With ibracket for magnetic strip for ML/2 autom. Unit recognition
TRUSTED_SUSTAINABILITY
Specifications
Product Type Lower Basket/Open Front
Model U 874/1
For Use With (Equipment) G 7804, G 7883, G7893, G7883CD, PG 8535 and PG 8536
Depth (Metric) 515 mm
Height (Metric) 50 mm
Width (Metric) 534 mm
Quantity 1 Each