missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX-1/OLR1 Antibody (214012) [Janelia Fluor™ 635], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB1564JF635
This item is not returnable.
View return policy
Description
LOX-1/OLR1 Monoclonal antibody specifically detects LOX-1/OLR1 in Mouse samples. It is validated for Flow Cytometry,Western Blot,CyTOF
Specifications
| LOX-1/OLR1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX-1, LOX1ox LDL receptor 1, LOXIN, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LOX-1, Arg60-Ile363, Accession # AAG44998 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Western Blot, CyTOF | |
| 214012 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Cardiovascular Biology, metabolism | |
| 4973 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction