missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2527CL1
This item is not returnable.
View return policy
Description
LOX Polyclonal antibody specifically detects LOX in Human,Mouse,Porcine,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Simple Western,KnockDown,Immunocytochemistry/Immunofluorescence
Specifications
| LOX | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Pig, Rat | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Simple Western, KnockDown | |
| CoraFluor 1 | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| A synthetic peptide made to an internal region of the human LOX protein (within residues 200-300). [Swiss-Prot P28300]. | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
| 4015 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction