Learn More
LOX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24877
This item is not returnable.
View return policy
Description
LOX Polyclonal specifically detects LOX in Human, Mouse, Rat, Porcine, Sheep samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockout Validated.
Specifications
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| LOX | |
| Human, Pig, Sheep | |
| Antisera |
| Polyclonal | |
| Unpurified | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |