Learn More
Invitrogen™ LOX Monoclonal Antibody (OTI9G6)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA525033
Product Code. 15984634
  • € 586.00 / 100µL
Description
LOX Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
Lysyl oxidase (LOX), a copper-containing amine oxidase, belongs to a heterogeneous family of enzymes that oxidize primary amine substrates to reactive aldehydes. It plays a vital role in the formation and repair of the extra cellular matrix. As well, LOX is a multifunctional enzyme having diverse biological functions such as developmental regulation, tumor suppression, cell motility and cellular senescence. The secreted form of LOX is responsible for the invasive properties of hypoxic human cancer cells. Thus, it is essential for hypoxia-induced metastasis and is a good therapeutic target for preventing and treating metastases.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LOX
Monoclonal
1.00 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P28300
LOX
Human recombinant protein fragment corresponding to amino acids 22-168 of LOX produced in HEK293T cell.
100 μL
Primary
Human
Antibody
IgG1
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
OTI9G6
Unconjugated
LOX
AI893619; cross-linking enzyme; H-rev142; LOX; Lysyl oxidase; Lysyl oxidase (an H-rev gene with its expression down-regulated in HRAS-transformed rat 208F fibroblasts); MGC105112; protein-lysine 6-oxidase; Protein-lysine 6-oxidase, long form; Protein-lysine 6-oxidase, short form; Ras excision protein; ras recision; rrg; Rrg1; TSC-160
Mouse
Affinity chromatography
RUO
4015
-20°C, Avoid Freeze/Thaw Cycles