Learn More
LOX Mouse anti-Human, Clone: OTI3C3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF500860
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is an extracellular copper enzyme that initiates the crosslinking of collagens and elastin. The enzyme catalyzes oxidative deamination of the epsilon-amino group in certain lysine and hydroxylysine residues of collagens and lysine residues of elastin. In addition to crosslinking extracellular matrix proteins, the encoded protein may have a role in tumor suppression.Specifications
| LOX | |
| Monoclonal | |
| Unconjugated | |
| LOX | |
| AAT10 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4015 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI3C3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P28300 | |
| LOX | |
| Human recombit protein fragment corresponding to amino acids 22-168 of human LOX produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.