missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX propeptide Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30327IR
This item is not returnable.
View return policy
Description
LOX propeptide Polyclonal antibody specifically detects LOX propeptide in Human, Rat samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| LOX propeptide | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, Simple Western, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| DyLight 755 | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot# P28300] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
| 4015 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction