missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX propeptide Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-41568H
This item is not returnable.
View return policy
Description
LOX propeptide Polyclonal antibody specifically detects LOX propeptide in Human, Mouse, Rat, Porcine samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot.
Specifications
| LOX propeptide | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody specifically recognizes both the pro-enzyme and pro-peptide forms of LOX protein in mouse and rat. | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoblot | |
| HRP | |
| PBS with No Preservative | |
| LOX | |
| A synthetic peptide made to an internal region of mouse LOX propeptide (residues 78-115). [UniProt# P28301] | |
| 0.1 mL | |
| Cancer, Cytoskeleton Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
| 4015 | |
| Human, Mouse, Rat, Pig, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction