missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOX propeptide Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-30327MFV500
This item is not returnable.
View return policy
Description
LOX propeptide Polyclonal antibody specifically detects LOX propeptide in Human, Rat samples. It is validated for Flow Cytometry
Specifications
| LOX propeptide | |
| Polyclonal | |
| Flow Cytometry | |
| lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase | |
| Synthetic peptide made to an internal portion of human LOX propeptide (within residues 118-168). [Swiss-Prot# P28300] | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Endothelial Cell Markers, Extracellular Matrix, Hypoxia, Signal Transduction | |
| 4015 | |
| Store at 4C in the dark. | |
| IgG |
| Flow Cytometry | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction