Learn More
Loxapine N-Oxide, Mikromol™
Description
Loxapine N-Oxide
Specifications
Specifications
| Chemical Name or Material | Loxapine N-Oxide |
| CAS | 25967-34-4 |
| Analyte or Component Names | Loxapine N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 Cl N3 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Dibenz[b,f][1,4]oxazepine, 2-chloro-11-(4-methyl-4-oxido-1-piperazinyl)-, Dibenz[b,f][1,4]oxazepine, 2-chloro-11-(4-methyl-1-piperazinyl)-, N-oxide (8CI), Cloxazepine N-oxide, Loxapine N-oxide |
| InChI Formula | InChI=1S/C18H18ClN3O2/c1-22(23)10-8-21(9-11-22)18-14-12-13(19)6-7-16(14)24-17-5-3-2-4-15(17)20-18/h2-7,12H,8-11H2,1H3 |
| SMILES | C[N+]1([O-])CCN(CC1)C2=Nc3ccccc3Oc4ccc(Cl)cc24 |
| IUPAC Name | 8-chloro-6-(4-methyl-4-oxidopiperazin-4-ium-1-yl)benzo[b][1,4]benzoxazepine |
| Show More |