Learn More
Loxapine.succinate, Lipomed
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Loxapine.succinate
Specifications
Specifications
| Chemical Name or Material | Loxapine Succinate |
| CAS | 27833-64-3 |
| Analyte or Component Names | Loxapine Succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 Cl N3 O . C4 H6 O4 |
| Synonym | Loxapine Succinate, 2-Chloro-11-(4-methyl-1-piperazinyl)dibenz[b,f][1,4 ]oxazepine butanedioate, Succinic acid, compd. with 2-chloro-11-(4-methyl-1-piperazinyl)dibenz[b,f][1,4 ]oxazepine, Dibenz[b,f][1,4 ]oxazepine, 2-chloro-11-(4-methyl-1-piperazinyl)-, butanedioate, 2-Chloro-11-(4-methyl-1-piperazinyl)-dibenz[b,f][1,4 ]oxazepine succinate, CL 71563, Daxolin, Loxapac, Loxitane, Loxitane Capsules |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CN1CCN(CC1)C2=Nc3ccccc3Oc4ccc(Cl)cc24.OC(=O)CCC(=O)O |
| IUPAC Name | butanedioic acid;8-chloro-6-(4-methylpiperazin-1-yl)benzo[b][1,4 ]benzoxazepine |
| Industry Type | Pharmaceutical |
| Show More |