missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Loxapine Succinate, LoGiCal
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Loxapine Succinate |
| CAS | 27833-64-3 |
| Analyte or Component Names | Loxapine Succinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H18 Cl N3 O . C4 H6 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Loxapine Succinate, 2-Chloro-11-(4-methyl-1-piperazinyl)dibenz[b,f][1,4]oxazepine butanedioate, Succinic acid, compd. with 2-chloro-11-(4-methyl-1-piperazinyl)dibenz[b,f][1,4]oxazepine, Dibenz[b,f][1,4]oxazepine, 2-chloro-11-(4-methyl-1-piperazinyl)-, butanedioate, 2-Chloro-11-(4-methyl-1-piperazinyl)-dibenz[b,f][1,4]oxazepine succinate, CL 71563, Daxolin, Loxapac, Loxitane, Loxitane Capsules |
| InChI Formula | InChI=1S/C18H18ClN3O.C4H6O4/c1-21-8-10-22(11-9-21)18-14-12-13(19)6-7-16(14)23-17-5-3-2-4-15(17)20-18;5-3(6)1-2-4(7)8/h2-7,12H,8-11H2,1H3;1-2H2,(H,5,6)(H,7,8) |
| Solution Type | Neat |
| SMILES | CN1CCN(CC1)C2=Nc3ccccc3Oc4ccc(Cl)cc24.OC(=O)CCC(=O)O |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction