Learn More
Loxapine succinate, MedChemExpress
Product Code. 16437276
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16437276
Brand: Medchem Express HY17390A
MedChemExpress Loxapine succinate is an orally active dopamine inhibitor, 5-HT receptor antagonist and also a dibenzoxazepine anti-psychotic agent.
Loxapine succinate is an orally active dopamine inhibitor, 5-HT receptor antagonist and also a dibenzoxazepine anti-psychotic agent.
Specifications
Chemical Name or Material Loxapine succinate
CAS 27833-64-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H24ClN3O5
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (224.27 mM; Need ultrasonic) ∣H2O : 2.5 mg/mL (5.61 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CCC(O)=O.CN1CCN(C2=NC3=CC=CC=C3OC4=CC=C(Cl)C=C24)CC1
Molecular Weight (g/mol) 445.9
Formula Weight 445.9
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only