missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LOXL1 Monoclonal Antibody (C1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA541635
This item is not returnable.
View return policy
Description
LOXL1 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.
Specifications
| LOXL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q08397 | |
| Loxl1 | |
| Recombinant protein Lysyl Oxidase Like Protein 1. The antigen corresponds to amino acid range 369-574 of the target protein. | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a κ |
| Immunohistochemistry (Paraffin), Western Blot | |
| C1 | |
| Unconjugated | |
| Loxl1 | |
| cross-linking enzyme; LOL; Lox2; LOXL; LOXL1; Lysyl oxidase 2; lysyl oxidase homolog 1; lysyl oxidase like 1; lysyl oxidase-like 1; lysyl oxidase-like protein 1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4016 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction