missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOXL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26608-1-AP-150UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the lysyl oxidase gene family. The prototypic member of the family is essential to the biogenesis of connective tissue, encoding an extracellular copper-dependent amine oxidase that catalyses the first step in the formation of crosslinks in collagens and elastin. A highly conserved amino acid sequence at the C-terminus end appears to be sufficient for amine oxidase activity, suggesting that each family member may retain this function. The N-terminus is poorly conserved and may impart additional roles in developmental regulation, senescence, tumor suppression, cell growth control, and chemotaxis to each member of the family.Specifications
| LOXL1 | |
| Polyclonal | |
| Unconjugated | |
| LOXL1 | |
| LOL, LOXL, LOXL1, Lysyl oxidase homolog 1, lysyl oxidase like 1, Lysyl oxidase like protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 4016 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q08397 | |
| LOXL1 | |
| LOXL1 Fusion Protein Ag24293 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction