Learn More
LOXL2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H7F1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67139-20UL
 View more versions of this product
Product Code. 18074505
  • € 145.00 / 20µL
Description
The LOXL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LOXL2. This antibody recognizes human antigen. The LOXL2 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
LOXL2
1H7F1
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG2a
Western Blot
1000 μg/mL
LOR2, LOXL2, Lysyl oxidase homolog 2, lysyl oxidase like 2, Lysyl oxidase like protein 2, WS9 14
20 μL
4017
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.