Learn More
LOXL2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H7F1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67139-100UL
Description
The LOXL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LOXL2. This antibody recognizes human antigen. The LOXL2 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| LOXL2 | |
| 1H7F1 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2a |
| Western Blot | |
| 1000 μg/mL | |
| LOR2, LOXL2, Lysyl oxidase homolog 2, lysyl oxidase like 2, Lysyl oxidase like protein 2, WS9 14 | |
| 100 μL | |
| 4017 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |