missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LOXL2 Recombinant Rabbit Monoclonal Antibody (23GB2610), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen MA553500
This item is not returnable.
View return policy
Description
LOXL2 Recombinant Monoclonal antibody specifically detects LOXL2 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
| LOXL2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LOXL2 | |
| 1110004B06Rik; 4930526G11Rik; 9430067E15Rik; LOR2; Loxl2; Lysyl oxidase homolog 2; lysyl oxidase homolog 2; LOW QUALITY PROTEIN: lysyl oxidase homolog 2; lysyl oxidase like 2; lysyl oxidase related 2; lysyl oxidase-like 2; lysyl oxidase-like 2 delta e13; lysyl oxidase-like 2 protein; lysyl oxidase-like protein 2; Lysyl oxidase-related protein 2; lysyl oxidase-related protein WS9-14; WS9-14 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 290350, 4017, 94352 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2610 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| B5DF27, P58022, Q9Y4K0 | |
| LOXL2 | |
| A synthesized peptide derived from human LOXL2 (711-774AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction