missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPAR1/LPA1/EDG-2 Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03363R
This item is not returnable.
View return policy
Description
LPAR1/LPA1/EDG-2 Polyclonal specifically detects LPAR1/LPA1/EDG-2 in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
| LPAR1/LPA1/EDG-2 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| edg-2, EDG2Lysophosphatidic acid receptor Edg-2, endothelial differentiation, lysophosphatidic acid G-protein-coupled receptor2,, Gpcr26, GPR26, LPA receptor 1, LPA-1, LPA1vzg-1, lysophosphatidic acid receptor 1, Mrec1.3, rec.1.3, ventricular zone gene 1, VZG1 | |
| Rabbit | |
| 43 kDa | |
| 0.1 mL | |
| Apoptosis, GPCR, Signal Transduction | |
| 1902 | |
| Store at 4C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| LPAR1 | |
| Synthetic peptide made to an internal portion of human EDG2 (within residues 200-300). [Swiss-Prot# Q92633] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat, Chicken | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction