Learn More
Invitrogen™ LPAR4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532902
Description
Percent identity with other species by BLAST analysis: Human, Gorilla (100%); Gibbon, Marmoset, Mouse, Rat, Bat, Hamster, Rabbit, Pig (94%); Monkey, Bovine, Panda, Horse (88%); Dog, Elephant (82%). For IHC(P), use heat induced antigen retrieval in pH 6.0 citrate buffer. After incubation with the primary antibody, slides were incubated with biotinylated secondary antibody, followed by alkaline phosphatase-streptavidin and chromogen.
EDG4 belongs to a family of G-protein coupled receptors whose ligands are lysophospholipids. There are eight known members of the EDG receptor family and they are implicated in mediating growth-related effects such as induction of cellular proliferation, alterations in differentiation and survival, and suppression of apoptosis. They also evoke cellular effector functions that are dependent on cytoskeletal responses such as contraction, secretion, adhesion and chemotaxis. EDG receptors are developmentally regulated and differ in tissue distribution. They couple to multiple types of G proteins to signal through ras and MAP kinase, rho, phospholipase C, and several protein tyrosine kinases. EDG4 is expressed in testes, ovarian tumor, and leukocyte containing tissues.
Specifications
| LPAR4 | |
| Polyclonal | |
| Unconjugated | |
| Lpar4 | |
| 5730485F04Rik; G protein-coupled receptor 23; Gpr23; G-protein coupled receptor 23; LPA receptor 4; LPA4; LPA-4; LPA4 Receptor; Lpar4; Lysophosphatidic acid receptor 4; P2RY9; P2Y purinoceptor 9; P2Y5-LIKE; P2Y5-like receptor; p2y9; Purinergic receptor 9; RGD1563686 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 2846 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q99677 | |
| Lpar4 | |
| Synthetic 17 amino acid peptide from C-terminal cytoplasmic domain of human LPAR4 / GPR23. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.