missing translation for 'onlineSavingsMsg'
Learn More

LPCAT3 Mouse Monoclonal Antibody, Proteintech

Mouse Monoclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen LPCAT3
Clone 1C1G3
Concentration 1000 μg/mL
Applications Western Blot, Immunohistochemistry, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30233912
View Documents
Proteintech
67882-1-IG-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30237992
View Documents
Proteintech
67882-1-IG-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The LPCAT3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LPCAT3. This antibody recognizes human, pig antigen. The LPCAT3 antibody has been validated for the following applications: WB, IHC, ELISA analysis.
Specifications

Specifications

LPCAT3
1000 μg/mL
Monoclonal
Liquid
RUO
10162
Recombinant Protein
Primary
Store at -20°C. Stable for one year after shipment.
1C1G3
Western Blot, Immunohistochemistry, ELISA
Unconjugated
Mouse
Human, Pig
C3F; MBOAT5; OACT5; nessy
IgG1
Protein A purification
Monoclonal Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.