missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LPCAT4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16639964 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16639964 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16639964 Supplier Invitrogen™ Supplier No. PA5103512

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total AGPAT7.

Members of the 1-acylglycerol-3-phosphate O-acyltransferase family, such as AGPAT7, catalyze the conversion of lysophosphatidic acid (LPA) to phosphatidic acid (PA), a precursor in the biosynthesis of all glycerolipids. Both LPA and PA are involved in signal transduction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen LPCAT4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LPCAT4
Gene Accession No. Q643R3
Gene Alias 1-acylglycerol-3-phosphate O-acyltransferase 7; 1-acylglycerol-3-phosphate O-acyltransferase 7 (lysophosphatidic acid acyltransferase, eta); 1-acylglycerophosphocholine O-acyltransferase; 1-acylglycerophosphoserine O-acyltransferase; 1-AGP acyltransferase 7; 1-AGPAT 7; 1-alkenylglycerophosphoethanolamine O-acyltransferase; 1-alkylglycerophosphocholine O-acetyltransferase; acyl-CoA:lysophosphatidylethanolamine acyltransferase 2; -acylglycerol-3-phosphate O-acyltransferase 7 (lysophosphatidic acid acyltransferase, eta); acyltransferase like 3; acyltransferase-like 3; AGPAT7; AI505034; Aytl3; LPAAT-eta; LPCAT4; LPEAT2; Lysophosphatidylcholine acyltransferase 4; lysophosphatidylethanolamine acyltransferase 2; Lysophospholipid acyltransferase LPCAT4; plasmalogen synthase; PLSC domain containing protein; RGD1311177
Gene Symbols LPCAT4
Host Species Rabbit
Immunogen A synthesized peptide derived from human LPCAT4(Accession Q643R3), corresponding to amino acid residues Y479-D524.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254531
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.