missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LPHN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533907
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey (100%) Marmoset, Mouse, Rat, Elephant, Horse, Rabbit, Opossum, Turkey, Platypus (94%).
The Latrophilin family of G protein-coupled receptors consists of three members, latrophilin-1, 2 and 3, each of which displays distinct tissue distribution and function. Latrophilin-1, the most characterized member of this family, acts as a receptor for a-latrotoxin, a component of venom from the black widow spider. Binding of a-latrotoxin to latrophilin-1 triggers synaptic vesicle exocytosis via both Ca2+-dependent and -independent mechanisms, resulting in vesicle depletion. Latrophilin-1 is abundant in brain and present in endocrine cells. Latrophilin-3 is also brain-specific, whereas latrophilin-2 expression is ubiquitous.
Specifications
| LPHN2 | |
| Polyclonal | |
| Unconjugated | |
| ADGRL2 | |
| Adgrl2; Adhesion G protein-coupled receptor L2; AI450192; Calcium-independent alpha-latrotoxin receptor 2; calcium-independent alpha-latrotoxin receptor homolog 2; Cirl2; Cirl-2; Cl2; Cl2aa; Cl2ac; Gm619; KIAA0786; latrophilin 1; latrophilin 2; latrophilin homolog 1; Latrophilin-2; LEC1; Lectomedin-1; LPHH1; Lphn2; mKIAA0786 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23266 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| O95490 | |
| ADGRL2 | |
| Synthetic 16 amino acid peptide from C-terminus of human LPHN2. | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction